missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-1 Antibody (7C8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 25 publications
252.00 € - 583.00 €
Specifications
| Antigen | Caveolin-1 |
|---|---|
| Clone | 7C8 |
| Concentration | 1.25 mg/mL |
| Dilution | Western Blot 1:1000-1:4000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1-2 ug/ 500 ug of protein, Immunohistochemistry-Paraffin 1:100-1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24758774), Immunoblotting reported in scientific literature (PMID 31759628) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18076955
|
Novus Biologicals
NB100-615SS |
0.025 mL |
252.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18759043
|
Novus Biologicals
NB100-615 |
0.1 mL |
583.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caveolin-1 Monoclonal specifically detects Caveolin-1 in Human, Mouse, Rat, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.Specifications
| Caveolin-1 | |
| 1.25 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| Unconjugated | |
| Mouse | |
| Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 | |
| CAV1 | |
| IgG2b | |
| Protein A purified | |
| This is specific for caveolin alpha and beta proteins. |
| 7C8 | |
| Western Blot 1:1000-1:4000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1-2 ug/ 500 ug of protein, Immunohistochemistry-Paraffin 1:100-1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24758774), Immunoblotting reported in scientific literature (PMID 31759628) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Sheep | |
| 857 | |
| Intracellular membrane protein-containing vesicles (containing GLUT4) from rat adipocytes | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 23 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title