missing translation for 'onlineSavingsMsg'
Learn More

Cav1.3 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Product Code. 18311996 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18311996 100 μg 100µL
18381817 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18311996 Supplier Novus Biologicals Supplier No. NBP315489100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Cav1.3 Polyclonal antibody specifically detects Cav1.3 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cav1.3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias CACN4calcium channel, neuroendocrine/brain-type, alpha 1 subunit, CACNL1A2voltage-dependent L-type calcium channel subunit alpha-1D, Calcium channel, L type, alpha-1 polypeptide, isoform 2, calcium channel, voltage-dependent, L type, alpha 1D subunit, Cav1.3, CCHL1A2voltage-gated calcium channel alpha 1 subunit, L type, alpha-1 polypeptide, voltage-gated calcium channel alpha subunit Cav1.3, Voltage-gated calcium channel subunit alpha Cav1.3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1882-2181 of human Cav1.3 (NP_000711.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 776
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.