Learn More
Fisherbrand™ Catalyst Platinum-Cured Silicone Formulation Stopped Tubing Assembly
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.6 mm
3.2 mm
4.8 mm
6.4 mm
8 mm
9.5 mm
Thickness (Metric):
1.6 mm
2.4 mm
3.2mm
4.8mm
6.4mm
Unit Size:
Pack of 12
Pack of 6
Specifications
Specifications
| Description | Stopped Tubing Assembly |
| Diameter (Metric) Inner | 0.8 mm |
| Thickness (Metric) | 1.6 mm |