Learn More
Behr Labor Technik X1000 Catalyst pellets (5,0 g K2SO4, 0,5 g CuSO4)
Brand:  Behr Labor Technik B00217934
X1000 Catalyst pellets (5,0 g K2SO4, 0,5 g CuSO4)
Product Code. 10417842
  • 604.80 € / Pack of 1000