Learn More
CataCXium A, TRC
Specifications
Specifications
| Chemical Name or Material | CataCXium A |
| CAS | 321921-71-5 |
| Molecular Formula | C24H39P |
| Quantity | 100 mg |
| Synonym | CXium A, Butylbis(tricyclo[3.3.1.13,7]dec-1-yl)phosphine, (n-Butyl)di(1-adamantyl)phosphine, Bis(adamant-1-yl)(butyl)phosphine, Bis(adamantan-1-yl)(butyl)phosphine, Butyldi(1-adamantanyl)phosphine, Butyldi(1-adamantyl)phosphineDi-1-adamantylbutylphosphine, Diadamantyl-n-butylphosphane |
| InChI Formula | InChI=1S/C24H39P/c1-2-3-4-25(23-11-17-5-18(12-23)7-19(6-17)13-23)24-14-20-8-21(15-24)10-22(9-20)16-24/h17-22H,2-16H2,1H3/t17-,18+,19-,20-,21+,22-,23?,24?,25? |
| SMILES | CCCCP(C12C[C@@H]3C[C@@H](C[C@@H](C3)C1)C2)C45C[C@@H]6C[C@@H](C[C@@H](C6)C4)C5 |
| IUPAC Name | bis(1-adamantyl)-butylphosphane |
| Molecular Weight (g/mol) | 358.54 |
| Formula Weight | 358.2789 |