missing translation for 'onlineSavingsMsg'
Learn More

Castor Oil, Carl Roth™

Product Code. 10295813
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
10295813 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10295813 Supplier Carl Roth Supplier No. 4702.1
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Specifications

Acidity ≤1.5mg KOH/g
CAS 8001-79-4
Absorbance ≤0.7
Melting Point 229°C
Color Colorless to Yellow
Density 0.952-0.965
Melting Range –10 to –18°C
Packaging Glass
Molecular Formula C57H104O9
Refractive Index 1.477 to 1.481
Quantity 1 L
Synonym castor oil, ricinus oil, olio di ricino, venelex, xenaderm, optase, trypsin complex, unii-d5340y2i9g, castor oil usp:jan, 1-o,2-o,3-o-tris z-12-hydroxy-9-octadecenoyl glycerol
InChI Key ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
Molecular Weight (g/mol) 933.45
PubChem CID 14030006
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.