missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASRegistryNumber1t Iron, Chromium, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
CASRegistryNumber1t Iron, Chromium, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Arsenic/Zinc/Zirconium/Lead/Titanium/Tin/Sulfur/Phosphorus/Niobium/Aluminium/Tungsten/Copper/Vanadium/Nickel/Molybdenum/Silicon/Manganese/Cobalt/Carbon/Chromium/Boron/Antimony/Oxygen/Nitrogen |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium -200 ppm/Arsenic -100 ppm/Zinc -90 ppm/Zirconium 0.0012 %/Lead 0.008 %/Titanium 0.008 %/Tin 0.026 %/Sulfur 0.048 %/Phosphorus 0.059 %/Niobium 0.091 %/Aluminium 0.122 %/Tungsten 0.202 %/Copper 0.213 %/Vanadium 0.27 %/Nickel 0.326 %/Molybdenum 0.363 %/Silicon 0.783 %/Manganese 1.02 %/Cobalt 1.55 %/Carbon 2.58 %/Chromium 28.5 %/Boron (<2.1) ppm/Antimony (<300) ppm/Oxygen (<40) ppm/Nitrogen (<600) ppm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction