missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAS Recombinant Rabbit Monoclonal Antibody (1W4T4)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA537829

Product Code. 17276703

  • 534.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Samples: HeLa, 293T, Jurkat, Mouse testis, Mouse thymus Immunogen sequence: LIGLFELPED DTIPDEEHFI DIEDTPGYQT AFSQLAFAGK KEHDPVGQMV NNPKIHLAQS LHKLSTACPG RVPSMVSTSL NAEALQYLQG YLQAASVTLL.

Proteins that carry a nuclear localization signal (NLS) are transported into the nucleus by the importin-alpha/beta heterodimer. Importin-alpha binds the NLS, while importin-beta mediates translocation through the nuclear pore complex. After translocation, RanGTP binds importin-beta and displaces importin-alpha. Importin-alpha must then be returned to the cytoplasm, leaving the NLS protein behind. The protein encoded by this gene binds strongly to NLS-free importin-alpha, and this binding is released in the cytoplasm by the combined action of RANBP1 and RANGAP1. In addition, the encoded protein may play a role both in apoptosis and in cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CAS
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P55060, Q9ERK4
CSE1L
A synthetic peptide corresponding to a sequence within amino acids 872-971 of human Exportin 2 (XPO2) (P55060).
100 μL
Primary
Human, Mouse
Antibody
IgG
ELISA, Western Blot
1W4T4
Unconjugated
CSE1L
2610100P18Rik; AA407533; Capts; CAS; Cellular apoptosis susceptibility protein; chromosome segregation 1 like; chromosome segregation 1-like (S. cerevisiae); chromosome segregation 1-like protein; CSE1; CSE1 chromosome segregation 1 like; CSE1 chromosome segregation 1-like; CSE1 chromosome segregation 1-like (yeast); CSE1L; Exp2; exportin-2; Importin-alpha re-exporter; MGC117283; MGC130036; MGC130037; XPO2
Rabbit
Affinity Chromatography
RUO
110750, 1434
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.