Learn More
Hecht Karl™ Cartridge
Product Code. 10013671
Click to view available options
Gas Type:
Butane
Unit Size:
Each
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10013671
Brand: Hecht Karl™ 41395001
Please to purchase this item. Need a web account? Register with us today!
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Specifications
Gas Type Butane
Size ∼450 g (approx. 880 mL)
compliance-icons
Product Identifier
  • butane gas cartridge, with safety valve, 450g
Signal Word
  • Danger
Hazard Category
  • Flammable gas.
Hazard Statement
  • H220-Extremely flammable gas.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.
  • P381-Eliminate all ignition sources if safe to do so.
  • P403-Store in a well-ventilated place.
Supplemental information
  • MIXTURE LIST-Contains : Hydrocarbons rich inC3-4