Learn More
Thermo Scientific™ CART (55-76), Rat
Product Code. p-5384620
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
Product Code. 15457609
Brand: Thermo Scientific™ J66304.!D
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For cellular and molecular biology applications
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 2440.81
Product Type CART (55-76), rat
Quantity 0.5 mg
Regulatory Status RUO - research use only
Sequence Ile-Pro-Ile-Tyr-Glu-Lys-Lys-Tyr-Gly-Gln-Val-Pro-Met-Cys-Asp-Ala-Gly-Glu-Gln-Cys-Ala-Val (Disulfide bridge: Cys68-Cys74)