Learn More
Carphenazine, TRC
Product Code. 17954145
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17954145 25 mg 25mg
17962535 50 mg 50mg
17935105 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17954145 Supplier Toronto Research Chemicals Supplier No. TRCC18430525MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2622-30-2
Molecular Formula C24 H31 N3 O2 S
Molecular Weight (g/mol) 425.59
Synonym 1-[10-[3-[4-(2-Hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-1-propanone Carfenazine, 1-Propanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl]- (6CI,7CI,8CI), Carfenazine, Carphenazin, Carphenazine, Proketazine
IUPAC Name 1-[10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazin-2-yl]propan-1-one
SMILES CCC(=O)c1ccc2Sc3ccccc3N(CCCN4CCN(CCO)CC4)c2c1
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C24H31N3O2S/c1-2-22(29)19-8-9-24-21(18-19)27(20-6-3-4-7-23(20)30-24)11-5-10-25-12-14-26(15-13-25)16-17-28/h3-4,6-9,18,28H,2,5,10-17H2,1H3
Formula Weight 425.2137
Chemical Name or Material Carphenazine