missing translation for 'onlineSavingsMsg'
Learn More

Carnosic acid, MedChemExpress

Product Code. 16414867
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16414867

Brand: Medchem Express HYN0644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Carnosic acid has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity.

Carnosic acid has demonstrated inhibition of oxidative stress and inflammation, suppression of cell proliferation, and antibacterial activity.

Specifications

Chemical Name or Material Carnosic acid
Physical Form Powder
CAS 3650-09-7
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H28O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 130 mg/mL (391.06 mM; Need ultrasonic)
SMILES OC([C@@]([C@@]1([H])CCC2=CC(C(C)C)=C3O)(CCCC1(C)C)C2=C3O)=O
Molecular Weight (g/mol) 332.43
Formula Weight 332.43
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.