missing translation for 'onlineSavingsMsg'
Learn More

CARM1-IN-1, MedChemExpress

Product Code. p-200099997
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478037

Brand: Medchem Express HY12759

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CARM1-IN-1 is a potent and specific CARM1(Coactivator-associated arginine methyltransferase 1) inhibitor with IC50 of 8.6 uM; shows very low activity against PRMT1 and SET7(IC50 > 600 uM).

CARM1-IN-1 is a potent and specific CARM1(Coactivator-associated arginine methyltransferase 1) inhibitor with IC50 of 8.6 uM

Specifications

Chemical Name or Material CARM1-IN-1
CAS 1020399-49-8
Color Orange
Purity Grade Notes Research
Molecular Formula C26H21Br2NO3
Quantity 5 mg
Solubility Information DMSO : ≥ 35 mg/mL (63.03 mM)
SMILES O=C1/C(CN(CC2=CC=CC=C2)C/C1=C\C3=CC=C(O)C(Br)=C3)=C/C4=CC=C(O)C(Br)=C4
Molecular Weight (g/mol) 555.26
Formula Weight 555.26
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.