Learn More
CARM1-IN-1, MedChemExpress
Product Code. 16425328
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16425328
Brand: Medchem Express HY12759
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress CARM1-IN-1 is a potent and specific CARM1(Coactivator-associated arginine methyltransferase 1) inhibitor with IC50 of 8.6 uM; shows very low activity against PRMT1 and SET7(IC50 > 600 uM).
CARM1-IN-1 is a potent and specific CARM1(Coactivator-associated arginine methyltransferase 1) inhibitor with IC50 of 8.6 uM
Specifications
Chemical Name or Material CARM1-IN-1
CAS 1020399-49-8
Color Orange
Purity Grade Notes Research
Molecular Formula C26H21Br2NO3
Quantity 10 mg
Solubility Information DMSO : ≥ 35 mg/mL (63.03 mM)
SMILES O=C1/C(CN(CC2=CC=CC=C2)C/C1=C\C3=CC=C(O)C(Br)=C3)=C/C4=CC=C(O)C(Br)=C4
Molecular Weight (g/mol) 555.26
Formula Weight 555.26
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death