Learn More
Carl Zeiss™ Replacement Bulbs for Microscopes and Fiber Optic Power Supplies
Product Code. 10217083
Change view
Click to view available options
Type:
Halogen Bulb
Halogen Reflector Bulb
Wattage:
20 w
30 w
150 w
For Use With (Equipment):
Axiostar Plus Student, Axiostar Plus ICS
Invertoskop™
KL1500 Fiber Optic Power Supply
KL200 Fiber Optic Power Supply
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Wattage For Use With (Equipment) Unit Size
10217083 Halogen Bulb 30 w Invertoskop™ Each
10569511 Halogen Reflector Bulb 20 w KL200 Fiber Optic Power Supply Each
10625113 Halogen Bulb 20 w Axiostar Plus Student, Axiostar Plus ICS Each
10469872 Halogen Bulb 150 w KL1500 Fiber Optic Power Supply Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Product Code. 10217083 Supplier Carl Zeiss™ Supplier No. 0000000402943
  • ONLINE EXCLUSIVE PRICE
    22.75 €
    22.00 € / Pack of 1
    Save 0.75 €
    3% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
For Carl Zeiss Microscopes and Fiber Optic Power Supplies
  • Accessory for use with Carl Zeiss Stereomicrscopes
  • Halogen bulb
Specifications
Type Halogen Bulb
For Use With (Equipment) Invertoskop™
Voltage 6 V
Wattage 30 w