Learn More
Carl Zeiss™ Lens Cleaning Tissues
Product Code. 10302075
Click to view available options
:
Lens Cleaning Tissue
This item is not returnable. View return policy
Product Code. 10302075
Brand: Carl Zeiss™ 4629750000000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For cleaning microscope lens and other optical surfaces
Recommended for:
  • Uncoated glass or polycarbonate lens surfaces
  • Television or computer display panels
  • Anti-reflection coated lens surfaces such as eyepieces or objective lenses
  • Mirror surfaces
  • Uncoated or coated glass or polymer filters
Specifications
Description Lens Cleaning Tissues
For Use With (Application) Cleaning microscope lens and other optical surfaces
Quantity 500/Pk.