missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carl Zeiss™ Goose-neck Light Guide
Click to view available options
Type:
Double Gooseneck Light Guide
Single Light Guide
Unit Size:
Each
Description
Compatible Microscopes:
- Stemi 2000
- Stemi 2000 C
- Stemi 2000 CS li>STEMI SV 11
- STEMI SV 6
Specifications
Specifications
| Type | Double Gooseneck Light Guide |
| Diameter (Metric) | 4.5 mm |
| For Use With (Equipment) | Zeiss Microscopes |
| Length (Metric) | 600 mm |
| Size | 4.5 x 600 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction