missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CARD11 Monoclonal Antibody (B.472.3)
GREENER_CHOICE

Product Code. 11585001
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11585001 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11585001 Supplier Invitrogen™ Supplier No. MA515037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein has a domain structure similar to that of CARD14 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD11
Applications Western Blot
Classification Monoclonal
Clone B.472.3
Concentration 344 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene CARD11
Gene Accession No. Q8CIS0, Q9BXL7
Gene Alias 0610008L17Rik; 2410011D02Rik; bcl10-interacting maguk protein 3; BENTA; BIMP3; Card11; CARD-containing MAGUK protein 1; Carma 1; CARMA1; caspase recruitment domain family member 11; caspase recruitment domain family, member 11; caspase recruitment domain-containing protein 11; IMD11; LOC100363332; MGC133069; PPBL
Gene Symbols CARD11
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding threonine 175 of CARD11.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108723, 84433
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.