missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase G2 (CPG2) Inhibitor, MedChemExpress

Product Code. 16414077
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16414077

Brand: Medchem Express HY70003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Carboxypeptidase G2 (CPG2) Inhibitor is a novel Carboxypeptidase G2 (CPG2) Inhibitor, Antitumor agents.

Carboxypeptidase G2 (CPG2) Inhibitor is a novel Carboxypeptidase G2 (CPG2) Inhibitor, Antitumor agents.

Specifications

Chemical Name or Material Carboxypeptidase G2 (CPG2) Inhibitor
CAS 192203-60-4
Color White
Purity Grade Notes Research
Molecular Formula C13H15NO6S
Quantity 50 mg
Synonym CPG2 Inhibitor
Solubility Information H2O : 1.43 mg/mL (4.56 mM; Need ultrasonic)
SMILES O=C(SC(C=C1)=CC=C1OC)N[C@@H](CCC(O)=O)C(O)=O
Molecular Weight (g/mol) 313.33
Formula Weight 313.33
Percent Purity 97.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.