Learn More
Novus Biologicals™ Carboxyl IR Fluorescent Particles, Aqua Green, 0.1-0.3 um
Product Code. 18614750 Shop All Bio Techne Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. 18614750
Brand: Novus Biologicals™ NBP318614
Please to purchase this item. Need a web account? Register with us today!
Aqua Green, 0.1-0.3 um
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C. Do not freeze.
Color Aqua Green
For Use With (Application) Flow Cytometry
Material Polystyrene
Product Type Fluroescent Carboxylate-Modified Particle
Quantity 10 mL
Diameter (Metric) 0.1 to 0.3 μm