Learn More
Carbostyril 124 N-Carboxymethyl Methanethiosulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | Carbostyril 124 N-Carboxymethyl Methanethiosulfonate |
| CAS | 1076199-73-9 |
| Molecular Formula | C13 H14 N2 O4 S2 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C13H14N2O4S2/c1-8-5-12(19-21(2,18)20)15-11-6-9(3-4-10(8)11)14-7-13(16)17/h3-6,14H,7H2,1-2H3,(H,16,17) |
| SMILES | Cc1cc(OS(=O)(=S)C)nc2cc(NCC(=O)O)ccc12 |
| IUPAC Name | 2-[(4-methyl-2-methylsulfonothioyloxyquinolin-7-yl)amino]acetic acid |
| Molecular Weight (g/mol) | 326.39 |
| Formula Weight | 326.0395 |