Learn More
Carbophenothion D10 (di(ethyl D5)) 100 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Carbophenothion D10 (di(ethyl D5)) |
| CAS | 2733387-54-5 |
| Solvent or Matrix | Acetone |
| Molecular Formula | C11 2H10 H6 Cl O2 P S3 |
| Quantity | 1 mL |
| Synonym | Carbophenothion D10 (di(ethyl D5)) |
| InChI Formula | InChI=1S/C11H16ClO2PS3/c1-3-13-15(16,14-4-2)18-9-17-11-7-5-10(12)6-8-11/h5-8H,3-4,9H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])OP(=S)(OC([2H])([2H])C([2H])([2H])[2H])SCSc1ccc(Cl)cc1 |
| IUPAC Name | (4-chlorophenyl)sulfanylmethylsulfanyl-bis(1,1,2,2,2-pentadeuterioethoxy)-sulfanylidene-λ^{5}-phosphane |
| Molecular Weight (g/mol) | 352.93 |
| Show More |