Learn More
Thermo Scientific™ Carbonate-buffered saline (5X), pH 9.5
Product Code. 15456089
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
Product Code. 15456089
Brand: Thermo Scientific™ J63928.AK
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5X
pH Range 9.5
Format Liquid
Quantity 250 mL
Storage Requirements Ambient temperatures
Type Carbonate-buffered saline