missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Resulfurized, AISI 1118, G11180, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, Resulfurized, AISI 1118, G11180, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Niobium/Titanium/Vanadium/Tin/Cobalt/Nitrogen/Phosphorus/Aluminium/Molybdenum/Chromium/Carbon/Copper/Nickel/Silicon/Manganese/Cadmium/Calcium/Zinc/Magnesium/Selenium/Antimony/Tantalum/Bismuth/Boron/Oxygen/Zirconium/Tungsten/Lead/Arsenic |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Niobium 0.0013 %/Titanium 0.003 %/Vanadium 0.003 %/Tin 0.01 %/Cobalt 0.0101 %/Nitrogen 0.011 %/Phosphorus 0.012 %/Aluminium 0.034 %/Molybdenum 0.045 %/Chromium 0.105 %/Carbon 0.162 %/Copper 0.191 %/Nickel 0.195 %/Silicon 0.3 %/Manganese 1.45 %/Cadmium (<0.0010) %/Calcium (<0.0010) %/Zinc (<0.0040) %/Magnesium (<0.0050) %/Selenium (<0.0050) %/Antimony (<0.0060) %/Tantalum (<0.0080) %/Bismuth (<0.0140) %/Boron (0.0005) %/Oxygen (0.004) %/Zirconium (0.004) %/Tungsten (0.005) %/Lead (0.006) %/Arsenic (0.008) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction