Learn More
Carbon Steel, Residuals, ARMI∣MBH
Product Code. 17967374
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17967374 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17967374 Supplier ARMI/MBH Supplier No. MBH12X 354 B
Please to purchase this item. Need a web account? Register with us today!
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Carbon Steel, Residuals, Ø38x20mm
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Nitrogen/Tin/Aluminium/Vanadium/Cobalt/Titanium/Tungsten/Arsenic/Molybdenum/Chromium/Phosphorus/Copper/Nickel/Niobium/Silicon/Carbon/Manganese
CAS 7439-89-6
Quantity 1 Ea.
Form Disc
Concentration or Composition (by Analyte or Components) Nitrogen 0.002 %/Tin 0.015 %/Aluminium 0.02 %/Vanadium 0.02 %/Cobalt 0.025 %/Titanium 0.025 %/Tungsten 0.025 %/Arsenic 0.03 %/Molybdenum 0.03 %/Chromium 0.05 %/Phosphorus 0.05 %/Copper 0.07 %/Nickel 0.08 %/Niobium 0.08 %/Silicon 0.2 %/Carbon 0.25 %/Manganese 5 %