missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1055, G10550, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Carbon Steel, AISI 1055, G10550, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Tin/Nitrogen/Arsenic/Molybdenum/Phosphorus/Nickel/Copper/Aluminium/Silicon/Chromium/Carbon/Manganese/Zinc |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tin 0.0018 %/Nitrogen 0.0051 %/Arsenic 0.0059 %/Molybdenum 0.0086 %/Phosphorus 0.0184 %/Nickel 0.0247 %/Copper 0.029 %/Aluminium 0.0325 %/Silicon 0.281 %/Chromium 0.338 %/Carbon 0.549 %/Manganese 0.685 %/Zinc (0.0016) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction