Learn More
(±)-Carbinoxamine-d6 Maleate (N,N-dimethyl-d6), CDN
Specifications
Specifications
| Chemical Name or Material | Carbinoxamine-d6 Maleate Salt |
| CAS | 2747914-08-3 |
| Molecular Formula | C16 D6 H13 Cl N2 O . C4 H4 O4 |
| Quantity | 0.01 g |
| Synonym | Carbinoxamine-d6 maleate, Carbinoxamine-D6 (N,N-dimethyl-D6) maleate, 2-[(4-Chlorophenyl)-pyridin-2-ylmethoxy]-N,N-bis(trideuteriomethyl)ethanamine (2 Z)-2-butenedioate, 2-[(4-Chlorophenyl)-2-pyridinylmethoxy]-N,N-di(methyl-d3)ethanamine (2 Z)-2-butenedioate, [2-[(4-Chlorophenyl)(pyridin-2-yl)methoxy]ethyl]di(methyl-d3)amine maleate |
| InChI Formula | InChI=1 S/C16H19ClN2O.C4H4O4/c1-19(2)11-12-20-16(15-5-3-4-10-18-15)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-10,16 H,11-12H2,1-2H3;1-2 H,(H,5,6)(H,7,8)/b;2-1-/i1D3,2D3; |
| SMILES | [2 H]C([2 H])([2 H])N(CCOC(c1ccc(Cl)cc1)c2ccccn2)C([2 H])([2 H])[2 H].OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;2-[(4-chlorophenyl)-pyridin-2-ylmethoxy]-N,N-bis(trideuteriomethyl)ethanamine |
| Molecular Weight (g/mol) | 412.897 |
| Formula Weight | 412.1672 g/mol |
| Show More |