missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG-Fc (Multi-species) Conjugate

Product Code. 18496758 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
This item is not returnable. View return policy

Product Code. 18496758

Brand: Thermo Scientific™ 7102852500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG-Fc (ms) Conjugate consists of a 13 kDa recombinant single domain antibody fragment that specifically binds to the Fc part of IgG from multiple species.

CaptureSelect Biotin Anti-IgG-Fc (ms) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of IgG from multiple species. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-IgG-Fc (ms) Conjugate allows you to:

  • Detect, quantitate, and characterize IgG antibodies from multiple species (all subclasses, IgG-specific), multi-species IgG-derived Fc fusion proteins, recombinant IgG antibodies, and total IgG in plasma and/or serum samples
  • Avoid cross-binding with other isotypes, such as IgA and IgM

Applications for CaptureSelect Biotin Anti-IgG-Fc (ms) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Content And Storage Store at -5°C to -30°C.
Form Liquid
Product Line CaptureSelect
Quantity 500 μg
Target IgG (Fc region)
Type Conjugate
Label or Dye Biotin & Other Haptens
Ligand Type Antibody
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.