Learn More
Captopril Disulfide, TRC
Product Code. 18001120
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18001120 5 mg 5mg
18021920 25 mg 25mg
18090010 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18001120 Supplier Toronto Research Chemicals Supplier No. TRCC1757605MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64806-05-9
Molecular Formula C18 H28 N2 O6 S2
Molecular Weight (g/mol) 432.55
Synonym 1,1'-[Disulfanediylbis-[(2S)-2-methyl-1-oxopropane-3,1-diyl]]-bis[(2S)-pyrrolidine-2-carboxylic] Acid, Captopril Imp. A (EP), Captopril Disulfide
IUPAC Name (2S)-1-[(2S)-3-[[(2S)-3-[(2S)-2-carboxypyrrolidin-1-yl]-2-methyl-3-oxopropyl]disulfanyl]-2-methylpropanoyl]pyrrolidine-2-carboxylic acid
SMILES C[C@H](CSSC[C@@H](C)C(=O)N1CCC[C@H]1C(=O)O)C(=O)N2CCC[C@H]2C(=O)O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C18H28N2O6S2/c1-11(15(21)19-7-3-5-13(19)17(23)24)9-27-28-10-12(2)16(22)20-8-4-6-14(20)18(25)26/h11-14H,3-10H2,1-2H3,(H,23,24)(H,25,26)/t11-,12-,13+,14+/m1/s1
Formula Weight 432.1389
Percent Purity >95%
Chemical Name or Material Captopril Disulfide
Grade HPLC