Learn More
Applied Biosystems™ CapSure™ LCM MicroCaps
Specially designed for use with the Arcturus™ Laser Capture Microdissection (LCM) instruments, CapSure™ Micro LCM Caps enable the precise and rapid extraction of pure populations of cells from heterogeneous sample preparations.
Specifications
| Product Type | LCM Cap |
|---|---|
| For Use With (Equipment) | ArcturusXT™ LCM Instrument |
| Shipping Condition | Room Temperature |
| Product Line | Arcturus™, CapSure™ |
Description
CapSure Micro LCM Caps may be used to isolate individual cells or several hundreds of cells on a single cap.
Key product features:
• Gentle application—non-damaging microdissection preserves the integrity of the captured material
• Versatile extraction—capture both large and small amounts of cells
Supports the integrity of microdissected cells for downstream molecular analysis
All CapSure LCM Caps have a proprietary transfer film bonded to the lower cap surface. When using the Arcturus microdissection systems an infrared laser pulses through the top of the cap during LCM and interacts with the transfer film, which then melts and bonds to the cells or regions of interest. The film absorbs the laser radiation—instead of the tissue or cell sample—creating a gentle, non-damaging microdissection that preserves the integrity of the captured material.
Go straight from capture to downstream analysis
CapSure Micro LCM Caps enable genetic material extraction with as little as 10 µL of lysis buffer. The new and improved design now enables the use of 0.2 mL microcentrifuge tubes. With this unique combination, you can proceed directly from LCM to downstream applications such as qPCR and next-generation sequencing with a single-tube workflow, while preserving precious sample.
Specifications
| LCM Cap | |
| ArcturusXT™ LCM Instrument | |
| Room Temperature | |
| Arcturus™, CapSure™ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.