missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPRIN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20766-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene may regulate the transport of mRNA. It may play a role in the differentiation of erythroblasts. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| CAPRIN2 | |
| Polyclonal | |
| Unconjugated | |
| CAPRIN2 | |
| C1QDC1, caprin 2, caprin family member 2, CAPRIN2, EEG 1, EEG1, KIAA1873, Protein EEG 1, RNA granule protein 140, RNG140 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 232560, 65981, 686779 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q05A80, Q6IMN6 | |
| CAPRIN2 | |
| CAPRIN2 Fusion Protein Ag14745 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction