Learn More
Thermo Scientific™ Thermo Scientific™ 90 mL Specimen Collection Kits
Description
The Thermo Scientific™ Urine Specimen Collection kits feature a flip-top 90mL collection vial and a specimen transport bag that are individually packaged for convenience. The kits are available with or without sealing tape for sample security.
Kit Components:
- 90 mL flip top container with twin valve design, and temperature strip
- Dual pocket specimen transport bag with absorbent pad
- 5 in x 8 in heat seal bag
- Evidence tape (Included in kit with SKU # 91020TV)
- Tamper Evident
- Lock Seal
- Individually bagged
Specifications
Specifications
| Product Type | Pain Management Specimen Kit |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Capacity (Metric) | 90 mL |
| Closure Type | Flip Top |
| Color | Natural |
| Description | Capitol Vial (90mL) Pain Management Specimen Kit in individual bags |
| Height (Metric) | 52 mm |
| No. per Case | 100/Cs. |