Learn More
Agilent Technologies CAPILLARY COLUMN CP-WAX 52 CB ULTIMETAL 50M X 0.53MM 2.00UM FILM THICKNESS EACH
Brand:  Agilent Technologies CP7179
Encompass_Preferred
CAPILLARY COLUMN CP-WAX 52 CB ULTIMETAL 50M X 0.53MM 2.00UM FILM THICKNESS EACH
Product Code. 10665631
  • 1844.04 € / Each