Learn More
Capecitabine-2',3'-cyclic Carbonate, TRC
Specifications
Specifications
| Chemical Name or Material | 2',3'-O-Carbonyl-5'-deoxy-5-fluoro-N4-(pentyloxycarbonyl)cytidine |
| CAS | 921769-65-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 H20 F N3 O7 |
| Quantity | 10 mg |
| Synonym | 2',3'-O-Carbonyl-5'-deoxy-5-fluoro-N4-(pentyloxycarbonyl)cytidine, Cytidine, 5'-deoxy-5-fluoro-N-[(pentyloxy)carbonyl]-, cyclic 2',3'-carbonate, Capecitabine-2',3'-cyclic carbonate |
| InChI Formula | InChI=1S/C16H20FN3O7/c1-3-4-5-6-24-15(22)19-12-9(17)7-20(14(21)18-12)13-11-10(8(2)25-13)26-16(23)27-11/h7-8,10-11,13H,3-6H2,1-2H3,(H,18,19,21,22)/t8-,10-,11-,13-/m1/s1 |
| SMILES | CCCCCOC(=O)NC1=NC(=O)N(C=C1F)[C@@H]2O[C@H](C)[C@H]3OC(=O)O[C@@H]23 |
| IUPAC Name | pentyl N-[1-[(3aR,4R,6R,6aR)-6-methyl-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-5-fluoro-2-oxopyrimidin-4-yl]carbamate |
| Molecular Weight (g/mol) | 385.34 |
| Show More |