missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Cap Strips

Product Code. 11982242
Change view
Click to view available options
:
Quantity: 288 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11982242 Quantity: 288 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11982242 Supplier Macherey-Nagel™ Supplier No. 740478.24
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Cap Strips for sealing of Tube Strips or Round‐well Blocks.

  • Provided in 8-well strip
  • For use with tube strips

Specifications

Format Strip Plate
No. of Wells 8
For Use With (Application) For Sealing Of Tube Strips
No. per Pack 288
Quantity 288/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.