missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Cap Strips for PCR 8-tube Strips

Product Code. 15975301
Change view
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
Unit Size:
Pack of 125
Pack of 1250
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15975301 Natural Pack of 1250
15985301 Red -
10073241 Red Pack of 125
10217161 Blue Pack of 125
15995301 Blue -
10469181 Green -
15905311 Green Pack of 1250
15915311 Yellow Pack of 1250
10499371 Yellow Pack of 125
10135201 Violet Pack of 125
15925311 Violet -
15935311 Assorted -
10517071 Assorted Pack of 125
10441872 Natural Pack of 125
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 15975301 Supplier Greiner Bio-One Supplier No. 373270

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with PCR 8-tube Strip. Greiner Bio-One Cap Strips are thin walled for optimal heat transfer.

  • Non-Pyrogenic
  • Domed lid
  • Highly transparent polypropylene
  • Free of detectable DNase, RNase, human DNA
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
  • Seals 8-tube strips in addition to
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens
Color Natural
Quantity 1250
Material Polypropylene
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.