missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Cap Strips for PCR 8-tube Strips
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Violet
Yellow
Unit Size:
Pack of 125
Pack of 1250
Description
- Non-Pyrogenic
- Domed lid
- Highly transparent polypropylene
- Free of detectable DNase, RNase, human DNA
- Thin wall for optimal heat transfer
- Low evaporation rate in PCR
- Seals 8-tube strips in addition to
Specifications
Specifications
| Color | Violet |
| Quantity | 1250/Cs. |
| For Use With (Equipment) | PCR 8-tube Strip |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Product Type | Cap Strip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction