Learn More
Canrenoic acid potassium salt, 98+%, Thermo Scientific™
66.40 € - 607.00 €
Chemical Identifiers
| CAS | 2181-04-6 |
|---|---|
| Molecular Formula | C19H19IN2S2 |
| Molecular Weight (g/mol) | 466.40 |
| MDL Number | MFCD05662375 |
| InChI Key | XWUVSDMFUQTKQC-UHFFFAOYSA-M |
| Synonym | 3,3'-diethylthiacyanine iodide, gnf-pf-5364, 3,3'-diethylthiacyanine iodide, dye content ∼97 %, 3,3 inverted exclamation marka-diethylthiacyanine iodide, 3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2e-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2z-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide |
| PubChem CID | 6504364 |
| IUPAC Name | (2Z)-3-ethyl-2-[(3-ethyl-1,3-benzothiazol-3-ium-2-yl)methylidene]-1,3-benzothiazole;iodide |
| SMILES | CCN1C2=CC=CC=C2SC1=CC3=[N+](C4=CC=CC=C4S3)CC.[I-] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15443999
|
Thermo Scientific Alfa Aesar
J61394.03 |
1g |
66.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15453999
|
Thermo Scientific Alfa Aesar
J61394.06 |
5g |
167.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15463999
|
Thermo Scientific Alfa Aesar
J61394.14 |
25g |
607.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 2181-04-6 | |
| 466.40 | |
| XWUVSDMFUQTKQC-UHFFFAOYSA-M | |
| 6504364 | |
| CCN1C2=CC=CC=C2SC1=CC3=[N+](C4=CC=CC=C4S3)CC.[I-] |
| C19H19IN2S2 | |
| MFCD05662375 | |
| 3,3'-diethylthiacyanine iodide, gnf-pf-5364, 3,3'-diethylthiacyanine iodide, dye content ∼97 %, 3,3 inverted exclamation marka-diethylthiacyanine iodide, 3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2e-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2z-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide | |
| (2Z)-3-ethyl-2-[(3-ethyl-1,3-benzothiazol-3-ium-2-yl)methylidene]-1,3-benzothiazole;iodide |
Specifications
| 2181-04-6 | |
| MFCD05662375 | |
| 3,3'-diethylthiacyanine iodide, gnf-pf-5364, 3,3'-diethylthiacyanine iodide, dye content ∼97 %, 3,3 inverted exclamation marka-diethylthiacyanine iodide, 3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2e-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide, 2z-3-ethyl-2-3-ethyl-1,3-benzothiazol-3-ium-2-yl methylidene-1,3-benzothiazole iodide | |
| CCN1C2=CC=CC=C2SC1=CC3=[N+](C4=CC=CC=C4S3)CC.[I-] | |
| 466.40 | |
| 396.58 | |
| Canrenoic acid potassium salt |
| C19H19IN2S2 | |
| 14,1750 | |
| XWUVSDMFUQTKQC-UHFFFAOYSA-M | |
| (2Z)-3-ethyl-2-[(3-ethyl-1,3-benzothiazol-3-ium-2-yl)methylidene]-1,3-benzothiazole;iodide | |
| 6504364 | |
| ≥98% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 218-554-9
missing translation for 'rtecsNumber' : TU3500000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures