missing translation for 'onlineSavingsMsg'
Learn More

Camostat mesylate, MedChemExpress

Product Code. p-200101411
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16496206

Brand: Medchem Express HY13512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Camostat mesylate (Camostat mesilate) is an orally active, synthetic serine protease inhibitor for chronic pancreatitis. Camostat mesylate, an inhibitor of TMPRSS2, shows antiviral activity against SARS-CoV-2. Camostat mesylate also inhibits the activity of prostasin, trypsin, and matriptase.

Camostat mesylate (Camostat mesilate) is an orally active, synthetic serine protease inhibitor for chronic pancreatitis. Camostat mesylate, an inhibitor of TMPRSS2, shows antiviral activity against SARS-CoV-2. Camostat mesylate also inhibits the activity of prostasin, trypsin, and matriptase.

Specifications

Chemical Name or Material Camostat mesylate
Physical Form Solid
CAS 59721-29-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H26N4O8S
For Use With (Application) Cancer-Kinase/protease
Synonym Camostat mesilate FOY305 FOY-S980
Solubility Information DMSO : 125 mg/mL (252.77 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (101.11 mM)
SMILES O=C(OCC(N(C)C)=O)CC1=CC=C(OC(C2=CC=C(NC(N)=N)C=C2)=O)C=C1.CS(=O)(O)=O
Molecular Weight (g/mol) 494.52
Formula Weight 494.52
Percent Purity 98.45%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.