missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CaMKIV Polyclonal antibody specifically detects CaMKIV in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
| Antigen | CaMKIV |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Tris-buffered Saline 0.1% BSA |
| Gene Alias | brain Ca(2+)-calmodulin-dependent protein kinase type IV, brain Ca++-calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase IV, calcium/calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase type IV catalytic chain, CAM kinase- GR, CAM kinase IV, CaM kinase-GR, caMK, CaMK IV, CaMK-GR, EC 2.7.11, EC 2.7.11.17, IV, MGC36771 |
| Host Species | Rabbit |
| Immunogen | Between 425 and C-term |
| Purification Method | Affinity purified |
| Quantity | 50 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?