Learn More
Honeywell KCL™ Camapren™ 720 Gloves
Description
- Excellent sensitivity
- Good grip when handling greasy and oily parts
- High flexibility at low temperatures
- High elasticity
- Extremely comfortable
Specifications
Specifications
| Latex Free | No |
| Material | Polychloroprene, Latex, Cotton |
| Size | 7 |
| Sterility | Non-sterile |
| Thickness (Metric) | 0.65 mm |
| Puncture Rating | 1 |
| Tear Rating | 1 |
| Length (Metric) | 290 to 310 mm |