Learn More
Calnexin (Endoplasmic Reticulum Marker) Mouse, Clone: CANX/1543, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 478.00 €
Specifications
Antigen Calnexin (Endoplasmic Reticulum Marker)
Clone CANX/1543
Concentration 200μg/mL
Applications ELISA, Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15961128
View Documents
EnquireBio
821-MSM3-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15971128
View Documents
EnquireBio
821-MSM3-P1
100 μg
ONLINE EXCLUSIVE PRICE
554.40 €
478.00 € / 100µg
Save 76.40 €
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Specifications
Calnexin (Endoplasmic Reticulum Marker)
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant N-terminal fragment of human Calnexin protein (apprx. aa 1-300)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
90kDa
CANX/1543
ELISA, Western Blot, Immunohistochemistry
Unconjugated
RUO
821
IgG1 κ
Protein A/G purified from Bioreactor concentrate.
Cytoplasmic
HeLa, MCF-7 or U2OS cells. Kidney or small intestine.