Learn More
Endecotts Calibration Ball for Sieve
Description
- Test sieve calibration samples supplied by Endecotts, provided with a Certificate of Calibration, fully traceable to National Standards, and supplied as glass calibration samples in single shot, single use, or single test vials for controlled and accurate sieve verification.
Specifications
Specifications
| Product Type | Calibration Sample |
| For Use With (Equipment) | For use with Sieves |
| Size | 2.8 mm |