missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calcyclin Monoclonal Antibody (7D11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515272
This item is not returnable.
View return policy
Description
MA5-15272 targets Calcyclin in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15272 immunogen is purified recombinant fragment of calcyclin expressed in E. Coli.
Calcyclin is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma.
Specifications
| Calcyclin | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.03% sodium azide | |
| Q9HB71 | |
| Cacybp | |
| Purified recombinant fragment of calcyclin expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 7D11 | |
| Unconjugated | |
| Cacybp | |
| 2A9; 5B10; CABP; CACY; CacyBP; Calcyclin; calcyclin binding protein; calcyclin-binding protein; GIG5; growth-inhibiting gene 5 protein; hCacyBP; PNAS-107; PRA; RP1-102G20.6; S100A6; S100A6-binding protein; S100A6BP; Siah-interacting protein; Siah-interacting protein (SIP); SIP | |
| Mouse | |
| Protein G | |
| RUO | |
| 27101 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction