Learn More
Lonza™ Calcium Chloride, 300 mM, 2 mL
Description
- Quality supplement for cell culture media.
- Included for use in select primary cell culture media kits.
- 1 Vial Containing 2 mL 300 mM Calcium Chloride
Specifications
Specifications
| Concentration | 300 mM |
| Product Type | Calcium Chloride |
| For Use With (Application) | Supplement for cell culture media |
| Quantity | 2 mL |