Learn More
Calcitriol, TRC
Product Code. 30041180
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041180 50 mg 50mg
18073150 10 mg 10mg
18093440 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30041180 Supplier Toronto Research Chemicals Supplier No. TRCC14450050MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32222-06-3
Molecular Formula C27 H44 O3
Molecular Weight (g/mol) 416.64
Synonym Calcitriol, (5Z,7E)-9,10-Secocholesta-5,7,10(19)-triene-1alpha,3beta,25-triol, 1,3-Cyclohexanediol, 4-methylene-5-[(2E)-2-[(1R,3aS,7aR)-octahydro-1-[(1R)-5-hydroxy-1,5-dimethylhexyl]-7a-methyl-4H-inden-4-ylidene]ethylidene]-, (1R,3S,5Z)-, 9,10-Secocholesta-5,7,10(19)-triene-1,3,25-triol, (1α,3β,5Z,7E)- (9CI), (1R,3S,5Z)-4-Methylene-5-[(2E)-2-[(1R,3aS,7aR)-octahydro-1-[(1R)-5-hydroxy-1,5-dimethylhexyl]-7a-methyl-4H-inden-4-ylidene]ethylidene]-1,3-cyclohexanediol, (3β,5Z,7E)-9,10-Secocholesta-5,7,10(19)-trienetriol, 1,25-Dihydroxycholecalciferol, 1,25-Dihydroxyvitamin D, 1,25-Dihydroxyvitamin D3, 1a,25-Dihydroxyvitamin D3, 1α,25-(OH)2D3, 1α,25-Dihydroxycholecalciferol, 1α,25-Dihydroxyvitamin D3, 1α,25-Hydroxyl-vitamin D3, Calcijex, DN 101, Dihydroxyvitamin D3, Panbonis, Ro 21-5535, Rocaltrol, Silkis, Solbone P, Soltriol, Topitriol, Toptriol
IUPAC Name (1R,3S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol
SMILES C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)C[C@H](O)C3=C)\CCC[C@]12C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1
Formula Weight 416.329
Percent Purity >95%
Chemical Name or Material Calcitriol
Grade HPLC