Learn More
Thermo Scientific Chemicals Calcitonin, human, 96%
Description
Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes. A carrier peptide that can be used to internalize fusion proteins
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Species | Human |
| Purification Method | HPLC |
| Purity | ≥97% |
| Common Name | Human C carcinoma |
| Content And Storage | -30°C to -10°C |
| Format | Powder |
| For Use With (Application) | Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes. A carrier peptide that can be used to internalize fusion proteins |
| Molecular Weight (g/mol) | 3417.90 |
| Product Type | Calcitonin, human, ≥97% |
| Quantity | 0.5 mg |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.