Learn More
DiscoverX CAL-101 (Idelalisib, GS-1101)
Click to view available options
:
10 mg
Description
CAL-101 (Idelalisib, GS-1101, Zydelig) is a synthetic drug that potently inhibits the phosphoinositide 3-kinase P110Δ(PI3KΔ), with 40-300-fold selectivity over p110α, β, and γ. CAL-101 is FDA approved for treatment of chronic lymphocytic leukemia (CLL) in combination with rituximab. Provided as 10mg of dry powder.
Specifications
Specifications
| Quantity | 10 mg |