missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CAGE1 Polyclonal Antibody

Product Code. 15902495
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15902495 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15902495 Supplier Invitrogen™ Supplier No. PA575964

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 96% (by SDS-PAGE).

CAGE1 (Cancer Antigen 1) is a Protein Coding gene. Diseases associated with CAGE1 include Fat Necrosis Of Breast and Amelogenesis Imperfecta, Type If. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAGE1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CAGE1
Gene Accession No. Q5IR70, Q66HB6, Q8TC20
Gene Alias 4933427I01Rik; bA69L16.7; CAGE1; CAGE-1; cancer antigen 1; cancer/testis antigen 3; Cancer/testis antigen 3 homolog; cancer/testis antigen 95; cancer/testis antigen gene 1; cancer-associated gene 1 protein; Cancer-associated gene 1 protein homolog; CT3; CT3 homolog; CT95; Ctag3
Gene Symbols CAGE1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 718-762 of Human CAGE1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285782, 306872, 71213
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.