Learn More
Caerulomycin A, MedChemExpress
Product Code. 16494838
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16494838
Brand: Medchem Express HY114495
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Caerulomycin A (Cerulomycin; Caerulomycin), an antifungal compound, induces generation of T cells, enhances TGF-β-Smad3 protein signaling via suppressing interferon-γ-induced STAT1 signaling. Antifungal and antibiotic activity, and used in autoimmune diseases.
Caerulomycin A (Cerulomycin
Specifications
Chemical Name or Material Caerulomycin A
Physical Form Solid
CAS 21802-37-9
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C12H11N3O2
For Use With (Application) COVID-19-immunoregulation
Synonym Cerulomycin Caerulomycin
Solubility Information DMSO : 150 mg/mL (654.36 mM; Need ultrasonic)
SMILES COC1=CC(C2=NC=CC=C2)=NC(/C=N/O)=C1.[(E)]
Molecular Weight (g/mol) 229.23
Formula Weight 229.23
Percent Purity 98.0%
Grade Research